Imprimer

Anti-IL6ST Mouse Monoclonal Antibody [clone: B-S12]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB27359-100EA 714 EUR
ABCAAB27359-100
Anti-IL6ST Mouse Monoclonal Antibody [clone: B-S12]
Anticorps
Type: Primary
Antigen: IL6ST (interleukin 6 signal transducer (gp130, oncostatin M receptor))
Clonality: monoclonal
Clone: B-S12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR