Imprimer

Anti-CD2 Mouse monoclonal antibody FITC (Fluorescein) [clone: B-E2]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB27392-1EA 241 EUR
ABCAAB27392-1
Anti-CD2 Mouse monoclonal antibody FITC (Fluorescein) [clone: B-E2]
Type: Primary
Antigen: CD2 (CD2 molecule)
Clonality: monoclonal
Clone: B-E2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR