Imprimer

Anti-CD40LG Mouse monoclonal antibody FITC (Fluorescein) [clone: B-B29]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB27407-1EA 241 EUR
ABCAAB27407-1
Anti-CD40LG Mouse monoclonal antibody FITC (Fluorescein) [clone: B-B29]
Type: Primary
Antigen: CD40LG (CD40 ligand)
Clonality: monoclonal
Clone: B-B29
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR