Imprimer

Anti-PSPHP1 Goat polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB27474-100EA 375 EUR
ABCAAB27474-100
Anti-PSPHP1 Goat polyclonal antibody
Type: Primary
Antigen: PSPHP1 (phosphoserine phosphatase pseudogene 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR