Imprimer

Anti-IVL Rabbit polyclonal antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB27495-100EA 696 EUR
ABCAAB27495-100
Anti-IVL Rabbit polyclonal antibody
Type: Primary
Antigen: IVL (involucrin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR