Imprimer

Anti-NR1H5P Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB48725-100EA 658 EUR
ABCAAB48725-100
Anti-NR1H5P Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: NR1H5P (nuclear receptor subfamily 1, group H, member 5, pseudogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR