Imprimer

Anti-ANXA5 Rabbit polyclonal antibody unconjugated

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB48815-100EA 673 EUR
ABCAAB48815-100
Anti-ANXA5 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: ANXA5 (annexin A5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR