Imprimer

Anti-AGTRAP Mouse Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB89247-50EA 878 EUR
ABCAAB89247-50
Anti-AGTRAP Mouse Polyclonal Antibody
Anticorps
Type: Primary
Antigen: AGTRAP (angiotensin II receptor-associated protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR