Imprimer
Anti-MYC Rabbit Polyclonal Antibody
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ABCAAB106952-100EA 628 EUR
ABCAAB106952-100
Anti-MYC Rabbit Polyclonal Antibody
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human