Imprimer

Anti-SNX1 Rabbit Monoclonal Antibody [clone: EPR7178]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB134126-100EA 780 EUR
ABCAAB134126-100
Anti-SNX1 Rabbit Monoclonal Antibody [clone: EPR7178]
Anticorps
Type: Primary
Antigen: SNX1 (sorting nexin 1)
Clonality: monoclonal
Clone: EPR7178
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR