Imprimer

Anti-Lactate Dehydrogenase Rabbit Monoclonal Antibody [clone: EP1563Y]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB134187-100
ABCAAB134187-100EA 899 EUR
ABCAAB134187-100
Anti-Lactate Dehydrogenase Rabbit Monoclonal Antibody [clone: EP1563Y]
Anticorps
Type: Primary
Antigen: Lactate Dehydrogenase
Clonality: monoclonal
Clone: EP1563Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR