Imprimer

Anti-SLC12A5 Mouse Monoclonal Antibody [clone: S1-12]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB134300-100
ABCAAB134300-100EA 720 EUR
ABCAAB134300-100
Anti-SLC12A5 Mouse Monoclonal Antibody [clone: S1-12]
Anticorps
Type: Primary
Antigen: SLC12A5 (solute carrier family 12 (potassium/chloride transporter), member 5)
Clonality: monoclonal
Clone: S1-12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR