Imprimer

Anti-CD68 Mouse Monoclonal Antibody (Biotin) [clone: Y1/82A]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB134352-50
ABCAAB134352-50EA 877 EUR
ABCAAB134352-50
Anti-CD68 Mouse Monoclonal Antibody (Biotin) [clone: Y1/82A]
Anticorps
Type: Primary
Antigen: CD68 (CD68 molecule)
Clonality: monoclonal
Clone: Y1/82A
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR