Imprimer

Anti-GLB1 Chicken Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB134435-200EA 835 EUR
ABCAAB134435-200
Anti-GLB1 Chicken Polyclonal Antibody
Anticorps
Type: Primary
Antigen: GLB1 (galactosidase, beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR