Imprimer

Anti-GPR34 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB134811-50
ABCAAB134811-50EA 838 EUR
ABCAAB134811-50
Anti-GPR34 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: GPR34 (G protein-coupled receptor 34)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR