Imprimer

Anti-TMP21 Rabbit Monoclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB134948-100
ABCAAB134948-100EA 820 EUR
ABCAAB134948-100
Anti-TMP21 Rabbit Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TMP21
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR