Imprimer

Anti-Brachyury / Bry Mouse Monoclonal Antibody [clone: 6C12]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB135254-100
ABCAAB135254-100EA 898 EUR
ABCAAB135254-100
Anti-Brachyury / Bry Mouse Monoclonal Antibody [clone: 6C12]
Anticorps
Type: Primary
Antigen: Brachyury / Bry
Clonality: monoclonal
Clone: 6C12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR