Imprimer

Anti-ITGAM Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: ICRF44]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB135929-50
ABCAAB135929-50EA 877 EUR
ABCAAB135929-50
Anti-ITGAM Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: ICRF44]
Anticorps
Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: monoclonal
Clone: ICRF44
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR