Imprimer

Anti-CEP97 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB136053-100
ABCAAB136053-100EA 838 EUR
ABCAAB136053-100
Anti-CEP97 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: CEP97 (centrosomal protein 97kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR