Imprimer

Anti-FAM35A+FAM35B Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB136079-100EA 798 EUR
ABCAAB136079-100
Anti-FAM35A+FAM35B Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: FAM35A+FAM35B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR