Imprimer

Anti-AVPR1B Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB136087-100
ABCAAB136087-100EA 838 EUR
ABCAAB136087-100
Anti-AVPR1B Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: AVPR1B (arginine vasopressin receptor 1B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR