Imprimer

Anti-HSD17B11 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB136109-100
ABCAAB136109-100EA 838 EUR
ABCAAB136109-100
Anti-HSD17B11 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: HSD17B11 (hydroxysteroid (17-beta) dehydrogenase 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR