Imprimer
Anti-ETFA Rabbit Monoclonal Antibody [clone: G22-L]
Fournisseur: Abcam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ABCAAB136615-500EA 853 EUR
ABCAAB136615-500
Anti-ETFA Rabbit Monoclonal Antibody [clone: G22-L]
Anticorps
Type: Primary
Antigen: ETFA (electron-transfer-flavoprotein, alpha polypeptide)
Clonality: monoclonal
Clone: G22-L
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human