Imprimer

Anti-EBV Latent Membrane Protein Rabbit Monoclonal Antibody [clone: D24-G]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB136633-250
ABCAAB136633-250EA 1060 EUR
ABCAAB136633-250
Anti-EBV Latent Membrane Protein Rabbit Monoclonal Antibody [clone: D24-G]
Anticorps
Type: Primary
Antigen: EBV Latent Membrane Protein
Clonality: monoclonal
Clone: D24-G
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR