Imprimer

Anti-NTN4 Goat Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB136663-50EA 798 EUR
ABCAAB136663-50
Anti-NTN4 Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: NTN4 (netrin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR