Imprimer

Anti-IgG Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB136765-500
ABCAAB136765-500EA 446 EUR
ABCAAB136765-500
Anti-IgG Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorps
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR