Imprimer
Anti-TNNC1 Rabbit Monoclonal Antibody
Fournisseur: Abcam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AB137130-100
ABCAAB137130-100EA 899 EUR
ABCAAB137130-100
Anti-TNNC1 Rabbit Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TNNC1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Commander maintenant