Imprimer

Anti-MARCH3 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB137261-100
ABCAAB137261-100EA 838 EUR
ABCAAB137261-100
Anti-MARCH3 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: MARCH3 (membrane-associated ring finger (C3HC4) 3, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR