Imprimer

Anti-CCR6 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB137369-100
ABCAAB137369-100EA 883 EUR
ABCAAB137369-100
Anti-CCR6 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: CCR6 (chemokine (C-C motif) receptor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR