Imprimer

Anti-VDR Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB137371-100
ABCAAB137371-100EA 849 EUR
ABCAAB137371-100
Anti-VDR Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: VDR (vitamin D (1,25- dihydroxyvitamin D3) receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR