Imprimer

Anti-E3 SUMO-protein ligase PIAS4 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB137500-100
ABCAAB137500-100EA 903 EUR
ABCAAB137500-100
Anti-E3 SUMO-protein ligase PIAS4 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: E3 SUMO-protein ligase PIAS4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR