Imprimer

Anti-ACSL4 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB137525-100
ABCAAB137525-100EA 907 EUR
ABCAAB137525-100
Anti-ACSL4 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: ACSL4 (acyl-CoA synthetase long-chain family member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR