Imprimer

Anti-SMYD5 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB137622-100
ABCAAB137622-100EA 882 EUR
ABCAAB137622-100
Anti-SMYD5 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: SMYD5 (SMYD family member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR