Imprimer

Anti-GRM3 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB137853-100
ABCAAB137853-100EA 883 EUR
ABCAAB137853-100
Anti-GRM3 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: GRM3 (glutamate receptor, metabotropic 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR