Imprimer

Anti-TAS2R14 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB138271-100
ABCAAB138271-100EA 878 EUR
ABCAAB138271-100
Anti-TAS2R14 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: TAS2R14 (taste receptor, type 2, member 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR