Imprimer

Anti-CTAG1B Mouse Monoclonal Antibody [clone: 4D8]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB139339-100
ABCAAB139339-100EA 893 EUR
ABCAAB139339-100
Anti-CTAG1B Mouse Monoclonal Antibody [clone: 4D8]
Anticorps
Type: Primary
Antigen: CTAG1B (cancer/testis antigen 1B)
Clonality: monoclonal
Clone: 4D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR