Imprimer

Anti-TFCP2L1 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB140197-100
ABCAAB140197-100EA 903 EUR
ABCAAB140197-100
Anti-TFCP2L1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: TFCP2L1 (transcription factor CP2-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR