Imprimer

Anti-MAP2K2 Mouse Monoclonal Antibody [clone: 1A2]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB140372-100EA 789 EUR
ABCAAB140372-100
Anti-MAP2K2 Mouse Monoclonal Antibody [clone: 1A2]
Type: Primary
Antigen: MAP2K2 (mitogen-activated protein kinase kinase 2)
Clonality: monoclonal
Clone: 1A2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR