Imprimer

Anti-PSMB2 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB140426-100
ABCAAB140426-100EA 883 EUR
ABCAAB140426-100
Anti-PSMB2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: PSMB2 (proteasome (prosome, macropain) subunit, beta type, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR