Imprimer

Anti-CDKN2AIP Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB140519-100EA 798 EUR
ABCAAB140519-100
Anti-CDKN2AIP Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: CDKN2AIP (CDKN2A interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR