Imprimer

Anti-SR Rabbit Monoclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB140602-100
ABCAAB140602-100EA 832 EUR
ABCAAB140602-100
Anti-SR Rabbit Monoclonal Antibody
Anticorps
Type: Primary
Antigen: SR
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR