Imprimer

Anti-LMO2 Mouse Monoclonal Antibody [clone: 4D8]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB140657-100EA 841 EUR
ABCAAB140657-100
Anti-LMO2 Mouse Monoclonal Antibody [clone: 4D8]
Anticorps
Type: Primary
Antigen: LMO2 (LIM domain only 2 (rhombotin-like 1))
Clonality: monoclonal
Clone: 4D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR