Imprimer

Anti-SUCNR1 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB140795-50
ABCAAB140795-50EA 883 EUR
ABCAAB140795-50
Anti-SUCNR1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: SUCNR1 (succinate receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: cytoplasmic domain
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR