Imprimer

Anti-P2RY14 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB140896-50
ABCAAB140896-50EA 838 EUR
ABCAAB140896-50
Anti-P2RY14 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: P2RY14 (purinergic receptor P2Y, G-protein coupled, 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR