Imprimer

Anti-SSTR2 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB140933-50EA 841 EUR
ABCAAB140933-50
Anti-SSTR2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: SSTR2 (somatostatin receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: extracellular domain
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR