Imprimer

Anti-PROKR1 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB140976-50
ABCAAB140976-50EA 798 EUR
ABCAAB140976-50
Anti-PROKR1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: PROKR1 (prokineticin receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR