Imprimer

Anti-ITGB1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 12G10]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB150002-50
ABCAAB150002-50EA 1030 EUR
ABCAAB150002-50
Anti-ITGB1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 12G10]
Anticorps
Type: Primary
Antigen: ITGB1 (integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12))
Clonality: monoclonal
Clone: 12G10
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR