Imprimer

Anti-TARBP1 Rabbit Monoclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB150412-100
ABCAAB150412-100EA 876 EUR
ABCAAB150412-100
Anti-TARBP1 Rabbit Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TARBP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR