Imprimer

Anti-TAAR1 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

AB150646-50
ABCAAB150646-50EA 878 EUR
ABCAAB150646-50
Anti-TAAR1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: TAAR1 (trace amine associated receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: extracellular domain
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR