Imprimer
Anti-IgG Goat Polyclonal Antibody (CF™ 660C)
Fournisseur: Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
20050-1EA 122 EUR
20050-1 20050.
Anti-IgG Goat Polyclonal Antibody (CF™ 660C)
Anticorps
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 660C Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse